D-ɑ-Tocopherol acetate solution 0.1 mCi/ml in ethanol, 25-50 mCi/mmol 0.925-1.85 GBq/mmol [ACETYL-1-14C]

Supplier: American Radiolabeled Chemicals

Synonyms: Vitamin E Acetate (D-form), (+)-ɑ-Tocopherol Acetate

ARC1323 10
102661-576EA 5463.38 USD
102661-576
D-ɑ-Tocopherol acetate solution 0.1 mCi/ml in ethanol, 25-50 mCi/mmol 0.925-1.85 GBq/mmol [ACETYL-1-14C]
D-alpha-Tocopherol acetate
Formula: C₃₁H₅₂O₃
MW: 472.75 g/mol
MDL Number: MFCD00072052
CAS Number: 58-95-7

Order Now

Specification Test Results

Specific Activity 25 - 50 mCi/mmol  (0.925-1.85 GBq/mmol)
Synonym Vitamin E acetate [1-14C]
Formula C31H52O3
Molecular Weight 472.76
Solvent E/E
Concentration 0.1 mCi/ml
Shipped in dry ice X

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR