2-Mercaptoethanol [1-¹⁴C] 0.1 mCi/ml in sterile water, 50-60 mCi/mmol 1.85-2.22 GBq/mmol

Supplier: American Radiolabeled Chemicals

ARC1128 250
102657-808EA 6448 USD
102657-808
2-Mercaptoethanol [1-¹⁴C] 0.1 mCi/ml in sterile water, 50-60 mCi/mmol 1.85-2.22 GBq/mmol
2-Mercaptoethanol [1-14C]
Formula: HSCH₂CH₂OH
MW: 78.13
CAS Number: 60-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR