Glutaryl-Coenzyme A 0.1 mCi/mL, [1,5-14C]

Supplier: American Radiolabeled Chemicals

Synonyms: CoA

ARC3471 25
102656-058EA 7420.31 USD
102656-058
Glutaryl-Coenzyme A 0.1 mCi/mL, [1,5-14C]
Glutary-Coenzyme A

  • Solvent: 0.01N NaOAc (pH 5.2) ethanol (1:1)
  • Specific activity: 1.85-2.22 GBq/mmol

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR