1,4-Dichlorobenzene [14C(U)] 0.1 mCi/ml in ethanol, 50-80 mCi/mmol 1.85-2.96 GBq/mmol

Supplier: American Radiolabeled Chemicals

ARC3218 100
102654-160EA 2694.15 USD
102654-160
1,4-Dichlorobenzene [14C(U)] 0.1 mCi/ml in ethanol, 50-80 mCi/mmol 1.85-2.96 GBq/mmol
1,4-Dichlorobenzene [14C(U)]
Formula: C₆H₄Cl₂
MW: 147.0 g/mol
CAS Number: 106-46-7

Order Now

Specification Test Results

Specific Activity 50 - 80 mCi/mmol  (1.85 - 2.96 GBq/mmol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR