tert-Butyl-5-amino-1H-indazole-1-carboxylate ≥98% (by HPLC)

17593-5G 17593-1G
102622-676EA 193.95 USD
102622-676 102642-188
tert-Butyl-5-amino-1H-indazole-1-carboxylate ≥98% (by HPLC)
tert-Butyl-5-amino-1H-indazole-1-carboxylate
Formula: C₁₂H₁₅N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD04114656
CAS Number: 129488-10-4

Order Now

Specification Test Results

Appearance Grey to Yellow or White Solid
¹H NMR Conforms to Structure
Purity by HPLC ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR