Cefonicid sodium 832 - 970 mcg/mg (by HPLC)

15160-5G
102633-088EA 109.62 USD
102633-088
Cefonicid sodium 832 - 970 mcg/mg (by HPLC)
Cefonicid sodium
Formula: C₁₈H₁₆N₆Na₂O₈S₃
Melting Pt: >160 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00941437
CAS Number: 61270-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR