Copper(II) acetylacetonate ≥98%
Supplier: Chem-Impex International
26722-1KG
26722-250G
26722-25G
26722-100G
26722-100G
102584-200EA
474.85
USD
102584-200
102603-948
102626-716
102626-718
CH26722-100G
Copper(II) acetylacetonate ≥98%
Copper (II) acetylacetonate
Formula:
C₁₀H₁₄CuO₄ |
MDL Number:
MFCD00000016 CAS Number: 13395-16-9 |
Specification Test Results
Molecular Weight | 261.76 |
Synonym | Cupric diacetylacetonate 2,4-Pentanedione copper(II) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...