D-Glucal ≥98%, powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol

21795-100G 21795-25G 21795-1G 21795-5G
102553-412EA 694.23 USD
102553-412 102563-732 102623-758 102634-390
D-Glucal ≥98%, powder
D-Glucal
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Melting Pt: 56…60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067186
CAS Number: 13265-84-4

Order Now

Specification Test Results

Heavy Metals ≤20 ppm
Loss on Drying ≤1%
Assay ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR