N1-Tritylpropane-1,3-diamineacetate ≥99% (by HPLC), white powder

07148-5G 07148-1G 07148-10G
102571-194EA 548.08 USD
102571-194 102620-364 102620-366
N1-Tritylpropane-1,3-diamineacetate ≥99% (by HPLC), white powder
N1-Tritylpropane-1,3-diamineacetate

Synonyms: Trt-NH(CH2)3NH2·AcOH

Formula: C₂₄H₂₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01631655
CAS Number: 325143-22-4

Order Now

Specification Test Results

Appearance White powder
Purity ≥ 99% (HPLC)
Melting Point 138-143 °C
Assay by titration ≥ 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR