Triethanolamine (trolamine) ≥99.5% (by GC)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

01718-1L
102616-432EA 51.6 USD
102616-432
Triethanolamine (trolamine) ≥99.5% (by GC)
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 190…193 °C (5 mmHg)
Density: 1.124 g/mL
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (by GC) ≥99.5%
Monoethanolamine ≤0.05%
Diethanolamine ≤0.40%
Water Content ≤0.2%
Color (APHA) ≤40
Refractive Index 1.4835

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR