Sodium amide ≥98% (by titrimetric analysis)

Supplier: Chem-Impex International

27268-100G 27268-100G
102604-518EA 146.08 USD
102604-518 CH27268-100G
Sodium amide ≥98% (by titrimetric analysis)
Sodium amide
Formula: H₂NNa
MDL Number: MFCD00011117
CAS Number: 7782-92-5
UN: 1390
ADR: 4.3,II

Order Now

Specification Test Results

Molecular Weight 39.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR