2,2,6,6-Tetramethylpiperidine ≥99%, liquid

Supplier: Chem-Impex International

24504-25G 24504-1KG 24504-250G 24504-100G 24504-100G 24504-250G 24504-25G
102601-562EA 27.08 USD
102601-562 102601-564 102613-650 102625-146 CH24504-100G CH24504-250G CH24504-25G
2,2,6,6-Tetramethylpiperidine ≥99%, liquid
2,2,6,6-Tetramethylpiperidine

2,2,6,6-Tetramethylpiperidine can be used as a hindered base (hindered amine) because it can dissolve in organic solvents unlike inorganic bases such as potassium hydroxide.


Synonym: Norpempidine

Formula: C₉H₁₉N
Boiling Pt: 152 – 153 °C
Density: 0,837 g/mL (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00005985
CAS Number: 768-66-1
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Refractive Index (n20/D) 1.445
Purity (GC) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR