1-Boc-piperidin-4-ylboronic acid pinacol ester ≥95%

Supplier: CHEM IMPEX INTL INC MS

Synonyms: tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, Piperidine-4-boronic acid, pinacol ester, N-BOC protected

19802-1G
102597-364EA 166.43 USD
102597-364
1-Boc-piperidin-4-ylboronic acid pinacol ester ≥95%
1-Boc-piperidin-4-ylboronic acid pinacol ester
Formula: C₁₆H₃₀BNO₄
MW: 311.2247 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09997792
CAS Number: 1048970-17-7

Order Now

Specification Test Results

Analysis by Mass Spectra Conforms to Structure
1H NMR Conforms to Structure
Purity ≥95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR