2-Bromethylamine hydrobromide 98-101% (by titrimetric analysis)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

14308-250G 14308-1KG 14308-25G 14308-100G 14308-25G
102594-364EA 117.97 USD
102594-364 102594-366 102621-456 102621-458 CH14308-25G
2-Bromethylamine hydrobromide 98-101% (by titrimetric analysis)
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Freezer
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now

Specification Test Results

Appearance White to off-white crystalline powder
Solubility Colorless solution (50 mg/ml in water)
Melting Point 170 - 175 °C
Water Content (Karl Fisher) ≤0.5%
pH 3 - 5
Infrared Spectrum Conforms to Structure
Assay by titration 98 - 101%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR