G418 disulfate

Supplier: Chem-Impex International

01417-1G 01417-5G 01417-25G 01417-1G 01417-5G
102573-590EA 58.46 USD
102573-590 102588-338 102588-340 CH01417-1G CH01417-5G
G418 disulfate
G418 disulphate

This geneticin disulfate is plant cell culture, tested.


  • Potency: ≥ 700 mg/mg (anhydrous)


White to off-white powder

Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR