FMOC isothiocyanate ≥95% (by NMR), pale yellow solid

Supplier: Chem-Impex International

12619-25G 12619-1G 12619-5G 12619-1G
102562-954EA 2417.94 USD
102562-954 102567-020 102630-156 CH12619-1G
FMOC isothiocyanate ≥95% (by NMR), pale yellow solid
FMOC isothiocyanate

Synonym: Fmoc-NCS

Formula: C₁₆H₁₁NO₂S
Melting Pt: 36…49 °C
Storage Temperature: Freezer
MDL Number: MFCD01861169
CAS Number: 199915-38-3

Order Now

Specification Test Results

Infrared Spectrum Conforms to Structure
13C NMR Conforms to Structure
1H NMR Conforms to Structure
Purity ≥95% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR