6-Aminocaproic acid 98.5-101.5%, white crystalline powder USP

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

02556-5KG 02556-250G 02556-1KG 02556-100G
102562-318EA 2095.26 USD
102562-318 102565-468 102589-400 102617-450
6-Aminocaproic acid 98.5-101.5%, white crystalline powder USP
6-Aminocaproic acid
Formula: C₆H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2

Order Now

Specification Test Results

Appearance White crystalline powder
Heavy Metals ≤20 ppm
Infrared Spectrum Conforms to Structure
Loss on Drying ≤0.5%
Residue On Ignition ≤0.1%
Assay 98.5 - 101.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR