(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥99% (by HPLC), white powder

04977-5G 04977-1G 04977-25G
102553-684EA 99.83 USD
102553-684 102556-450 102591-308
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥99% (by HPLC), white powder
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid
Formula: C₁₆H₂₂N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00237342
CAS Number: 16947-84-5

Order Now

Specification Test Results

Optical Rotation, [a]D²⁵ –12.0 ±1° (C=1 in MeOH)
¹³C NMR Conforms to Structure
¹H NMR Conforms to Structure
Purity by HPLC ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR