1,2-Epoxycyclopentane ≥95%
Supplier: Matrix Scientific
075520-5G
075520-25G
075520-100G
102541-730EA
52.31
USD
102541-730
102551-966
102551-968
1,2-Epoxycyclopentane ≥95%
1,2-Epoxycyclopentane
Hazard: Flammable/Irritant
Formula:
C₅H₈O Boiling Pt: 101…102 °C Density: 0.965 Flash Pt: 10 °C |
MDL Number:
MFCD00005161 CAS Number: 285-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...