5-Nitro-o-toluidine ≥95%

Supplier: Matrix Scientific

074974-100G 074974-500G
102539-126EA 49.23 USD
102539-126 102551-930
5-Nitro-o-toluidine ≥95%
5-Nitro-o-toluidine

Hazard: Toxic

Formula: C₇H₈N₂O₂
Melting Pt: 105…108 °C
MDL Number: MFCD00007741
CAS Number: 99-55-8
UN: 2660
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR