Methyl-1H-indazole-5-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 1H-indazole-5-carboxylate

069834-25G
102551-760EA 152.31 USD
102551-760
Methyl-1H-indazole-5-carboxylate ≥97%
Methyl 1H-indazole-5-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD07371609
CAS Number: 473416-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR