Daidzein

Supplier: Matrix Scientific

Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one

065243-1G
102551-676EA 235.38 USD
102551-676
Daidzein
Daidzein

Hazard: Irritant

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Boiling Pt: 513
MDL Number: MFCD00016954
CAS Number: 486-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR