2-Adamantanol

Supplier: Matrix Scientific

063697-5G 063697-25G
102551-632EA 36.92 USD
102551-632 102551-634
2-Adamantanol
2-Adamantanol

Hazard: Irritant

Formula: C₁₀H₁₆O
MDL Number: MFCD00074744
CAS Number: 700-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR