Benzo[b]thiophene-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-Benzothiophene-3-carboxylic acid

010137-1G
102550-950EA 33.85 USD
102550-950
Benzo[b]thiophene-3-carboxylic acid
Benzo[b]thiophene-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₆O₂S
Boiling Pt: 305…309 °C
Melting Pt: 174…175 °C
MDL Number: MFCD01846406
CAS Number: 5381-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR