(±)-3-Fluoro-α-methylbenzylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3-Fluorophenyl)ethanamine

008969-1G 008969-5G
102550-932EA 84.62 USD
102550-932 102550-934
(±)-3-Fluoro-α-methylbenzylamine ≥97%
(±)-3-Fluoro-α-methylbenzylamine

Hazard: Irritant/Refrigerate

Formula: C₈H₁₀FN
MW: 139.17 g/mol
Density: 1.059
MDL Number: MFCD04038308
CAS Number: 74788-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR