3-Chloro-2-fluorobenzyl alcohol
Supplier: Matrix Scientific
002096-5G
002096-25G
102550-810EA
181.54
USD
102550-810
102550-812
3-Chloro-2-fluorobenzyl alcohol
3-Chloro-2-fluorobenzyl alcohol
Hazard: Irritant
Formula:
C₇H₆ClFO Melting Pt: 39…41 °C |
MDL Number:
MFCD01631549 CAS Number: 261723-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...