Imidazolidin-4-one hydrochloride ≥95%
Supplier: Matrix Scientific
076581-250MG
076581-1G
076581-5G
102546-944EA
400
USD
102546-944
102546-946
102546-948
Imidazolidin-4-one hydrochloride ≥95%
Imidazolidin-4-one hydrochloride
Hazard: Irritant
Formula:
C₃H₇ClN₂O |
MDL Number:
MFCD22194318 CAS Number: 1373253-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...