(1-Methyl-1H-pyrazol-4-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

076570-250MG 076570-1G
102546-888EA 516.92 USD
102546-888 102546-890
(1-Methyl-1H-pyrazol-4-yl)methanamine hydrochloride ≥95%
(1-Methyl-1H-pyrazol-4-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClN₃
MW: 147.61 g/mol
MDL Number: MFCD11042229
CAS Number: 1107601-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR