2-((6-Chloropyridazin-3-yl)methyl)isoindoline-1,3-dione ≥95%

Supplier: Matrix Scientific

076567-250MG 076567-1G 076567-5G
102546-870EA 640 USD
102546-870 102546-872 102546-874
2-((6-Chloropyridazin-3-yl)methyl)isoindoline-1,3-dione ≥95%
2-((6-Chloropyridazin-3-yl)methyl)isoindoline-1,3-dione

Hazard: Irritant

Formula: C₁₃H₈ClN₃O₂
MW: 273.68 g/mol
MDL Number: MFCD14706915
CAS Number: 948996-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR