Cyclopropylméthanol ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclopropyl carbinol, Cyclopropanemethanol

076553-100G
102546-806EA 127.69 USD
102546-806
Cyclopropylméthanol ≥95%
Cyclopropylméthanol

Hazard: Flammable

Formula: C₄H₈O
Boiling Pt: 123…124 °C
Density: 0.904
Flash Pt: 35 °C
MDL Number: MFCD00001309
CAS Number: 2516-33-8
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR