5-Amino-N-ethyl-2-methyl-N-phenylbenzenesulfonamide ≥95%

Supplier: Matrix Scientific

076438-5G 076438-10G 076438-100G
102546-186EA 143.08 USD
102546-186 102546-188 102546-190
5-Amino-N-ethyl-2-methyl-N-phenylbenzenesulfonamide ≥95%
5-Amino-N-ethyl-2-methyl-N-phenylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂S
MW: 290.3800 g/mol
MDL Number: MFCD00025373
CAS Number: 51123-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR