(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-tert-Butyl 2-Methyl (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, (2S,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylic Acid 1-tert-Butyl 2-Methyl Ester, N-Boc-cis-4-hydroxy-L-proline Methyl Ester, Boc-cis-Hyp-OMe
076435-1G
076435-5G
102546-170EA
33.85
USD
102546-170
102546-172
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate ≥95%
(2S,4S)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₉NO₅ |
MDL Number:
MFCD00237541 CAS Number: 102195-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...