trans-4-(Boc-amino)cyclohexanemethylamine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (trans-4-(aminomethyl)cyclohexyl)carbamate

076419-250MG 076419-1G 076419-5G
102546-088EA 509.23 USD
102546-088 102546-090 102546-092
trans-4-(Boc-amino)cyclohexanemethylamine ≥95%
trans-4-(Boc-amino)cyclohexanemethylamine

Hazard: Irritant

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD01861810
CAS Number: 177583-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR