2,2'-Bipyridyl ≥95%

Supplier: Matrix Scientific

076381-100G
102545-880EA 75.38 USD
102545-880
2,2'-Bipyridyl ≥95%
2,2'-Bipyridyl

Hazard: Irritant

Formula: C₁₀H₈N₂
MDL Number: MFCD00006212
CAS Number: 366-18-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86bbc29edb6c5d09c058b8efcb6b9eb7f4d070c&vid=9e3a03e37045643109ae0765802e1997d30604ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR