3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-4-(3-fluorobenzyloxy)aniline

076336-1G 076336-5G
102545-640EA 46.15 USD
102545-640 102545-642
3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥95%
3-Chloro-4-((3-fluorobenzyl)oxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₁ClFNO
MDL Number: MFCD06809822
CAS Number: 202197-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR