Diethyl (2-oxopropyl)phosphonate ≥95%

Supplier: Matrix Scientific

Synonyms: Diethyl acetylmethylphosphonate

076304-5G
102545-482EA 76.92 USD
102545-482
Diethyl (2-oxopropyl)phosphonate ≥95%
Diethyl (2-oxopropyl)phosphonate

Hazard: Irritant

Formula: C₇H₁₅O₄P
Density: 1.01
MDL Number: MFCD00044728
CAS Number: 1067-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR