7-Amino-4,5-dihydro-5-oxopyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Amino-4,5-dihydro-5-oxo-pyrazolo[1,5-a]pyrimidine-3-carbonitrile

076229-250MG 076229-1G 076229-5G
102545-096EA 498.46 USD
102545-096 102545-098 102545-100
7-Amino-4,5-dihydro-5-oxopyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
7-Amino-4,5-dihydro-5-oxopyrazolo[1,5-a]pyrimidine-3-carbonitrile

Hazard: Irritant

Formula: C₇H₅N₅O
MDL Number: MFCD11040171
CAS Number: 138904-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR