Luteolin ≥95%

Supplier: Matrix Scientific

076178-1G 076178-5G 076178-10G
102544-850EA 38.46 USD
102544-850 102544-852 102544-854
Luteolin ≥95%

Hazard: Irritant

Formula: C₁₅H₁₀O₆
MDL Number: MFCD00017309
CAS Number: 491-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR