Ethyl indol-3-ylacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-indoleacetate, Ethyl 2-(1H-indol-3-yl)acetate, Ethyl 3-Indoleacetate, IAA ethyl ester, Indole-3-acetic acid ethyl ester, Ethyl indole-3-acetate, Indol-3-ylacetic acid ethyl ester, 3-Indoleacetic acid ethyl ester

076171-5G 076171-10G 076171-100G
102544-814EA 46.15 USD
102544-814 102544-816 102544-818
Ethyl indol-3-ylacetate ≥95%
Ethyl indol-3-ylacetate

Hazard: Irritant

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD00005635
CAS Number: 778-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR