Chromone-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid

076164-1G 076164-5G 076164-10G
102544-778EA 233.85 USD
102544-778 102544-780 102544-782
Chromone-2-carboxylic acid ≥95%
Chromone-2-carboxylic Acid

Hazard: Irritant

Formula: C₁₀H₆O₄
MDL Number: MFCD00006838
CAS Number: 4940-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR