L-Methionine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

076155-100G
102544-740EA 78.46 USD
102544-740
L-Methionine ≥95%
L-Methionine

Hazard: Irritant

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD00063097
CAS Number: 63-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR