DL-Lysine monohydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride

076147-10G 076147-100G
102544-704EA 36.92 USD
102544-704 102544-706
DL-Lysine monohydrochloride ≥95%
DL-Lysine monohydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O₂
MDL Number: MFCD00064563
CAS Number: 70-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR