Cephalothin ≥95%
Supplier: Matrix Scientific
Synonyms:
(6R,7R)-3-(Acetoxymethyl)-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(2-Thienylacetamido)cephalosporanic Acid, Cephalotin
076118-1G
076118-5G
102544-576EA
38.46
USD
102544-576
102544-578
Cephalothin ≥95%
Cephalothin
Purity: 95+%
CAS: 153-61-7
MDL Number: MFCD00242614
Molecular Formula: C16H16N2O6S2
Molecular Weight: 396,44
Hazard: Irritant
Formula:
C₁₆H₁₆N₂O₆S₂ |
MDL Number:
MFCD00242614 CAS Number: 153-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...