4-(1-Hydroxy-2-(methylamino)ethyl)phenol hydrochloride ≥95%
Supplier: Matrix Scientific
076117-10G
076117-100G
102544-572EA
107.69
USD
102544-572
102544-574
4-(1-Hydroxy-2-(methylamino)ethyl)phenol hydrochloride ≥95%
4-(1-Hydroxy-2-(methylamino)ethyl)phenol hydrochloride
Hazard: Irritant
Formula:
C₉H₁₄ClNO₂ |
MDL Number:
MFCD00050566 CAS Number: 5985-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...