(±)-α-Lipoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid

076109-10G
102544-536EA 364.62 USD
102544-536
(±)-α-Lipoic acid ≥95%
(±)-α-Lipoic acid

Hazard: Irritant

Formula: C₈H₁₄O₂S₂
MW: 206.33 g/mol
Boiling Pt: 160…165 °C
Melting Pt: 59…63 °C
MDL Number: MFCD00005474
CAS Number: 1077-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR