4-Cyano-3-methylbenzoic acid ≥95%

Supplier: Matrix Scientific

075917-1G 075917-5G 075917-10G
102543-620EA 178.46 USD
102543-620 102543-622 102543-624
4-Cyano-3-methylbenzoic acid ≥95%
4-Cyano-3-methylbenzoic acid

Hazard: Irritant

Formula: C₉H₇NO₂
MDL Number: MFCD14582961
CAS Number: 73831-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR