1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one ≥95%

Supplier: Matrix Scientific

075777-100G
102542-970EA 138.46 USD
102542-970
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one ≥95%
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one

Hazard: Irritant

Formula: C₅H₄N₄O
MDL Number: MFCD00599413
CAS Number: 315-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR