Minoxidil ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten

075760-100G
102542-896EA 553.85 USD
102542-896
Minoxidil ≥95%
Minoxidil

Hazard: Irritant

Formula: C₉H₁₅N₅O
MW: 209.253 g/mol
MDL Number: MFCD00063409
CAS Number: 38304-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR