(R)-(+)-1-(1-Naphthyl)ethylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

075741-5G
102542-804EA 30.77 USD
102542-804
(R)-(+)-1-(1-Naphthyl)ethylamine ≥95%
(R)-(+)-1-(1-Naphthyl)ethylamine

Hazard: Corrosive/Air Sensitive

Formula: C₁₂H₁₃N
Boiling Pt: 152…153 °C (11 mmHg)
Density: 1.06
MDL Number: MFCD00064114
CAS Number: 3886-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR