Terephthalonitrile ≥95%

Supplier: Matrix Scientific

075734-100G
102542-770EA 126.15 USD
102542-770
Terephthalonitrile ≥95%
Terephthalonitrile

Hazard: Irritant

Formula: C₈H₄N₂
Melting Pt: 224…227 °C
MDL Number: MFCD00001810
CAS Number: 623-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR